siRNA Details
Gene Symbol: Tnfsf13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159505.1 | NP_001152977.1 | 2 | 584 | ||
NM_023517.2 | NP_076006.2 | 2 | 584 | |||
GenBank mRNA | AK009514.1 | - | 2 | 586 | ||
AK040667.1 | - | 2 | 362 | |||
AK134028.1 | - | 2 | 290 | |||
AK170351.1 | - | 2 | 591 | |||
BC069900.1 | - | 2 | 543 | 5290965 |
Gene Symbol: Tnfsfm13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034097.2 | NP_001029269.1 | 9 | 1140 | ||
NM_001159503.2 | NP_001152975.1 | 9 | 1137 | |||
GenBank mRNA | AK044387.1 | - | 9 | 1141 | ||
BC096441.1 | - | 9 | 849 | 3482756 |

More Information
Other Species Targeted

Gene Ontology Categories:
