siRNA Details
Gene Symbol: Rsc1a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023544.5 | NP_076033.4 | 1 | 293 | ||
GenBank mRNA | AI854132.1 | - | 1 | 340 | ||
BC150752.1 | - | 1 | 293 | 9087663 |
Gene Symbol: Ddi2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017966.2 | NP_001017966.1 | 10 | 1678 | ||
GenBank mRNA | AK129015.1 | - | 1657 | |||
BC021415.1 | - | 10 | 1659 | |||
BC031491.1 | - | 3 | 432 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001034149, XM_006239308, XM_006239309, XM_006239306, XM_006239305, and others.
Gene Ontology Categories:
