siRNA Details
Gene Symbol: Pycard | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023258.4 | NP_075747.3 | 3 | 695 | ||
GenBank mRNA | AB032249.1 | - | 3 | 567 | ||
AF310104.1 | - | 3 | 507 | |||
AK007742.1 | - | 3 | 565 | |||
AK009852.1 | - | 3 | 570 | |||
AK152677.1 | - | 3 | 555 | |||
BC008252.1 | - | 3 | 537 | 2648391 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
