siRNA Details
Gene Symbol: Krt42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_212483.2 | NP_997648.2 | 8 | 1307 | ||
GenBank mRNA | AK019115.1 | - | 5 | 824 | ||
AK019133.1 | - | 5 | 769 | |||
AY459292.1 | - | 8 | 1307 | |||
BC112371.1 | - | 8 | 1299 | |||
BC148204.1 | - | 8 | 1307 | 40056537 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
