siRNA Details
Gene Symbol: Tnfsf13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159505.1 | NP_001152977.1 | 3, 4 | 660 | ||
GenBank mRNA | AK040667.1 | - | 3, 4 | 438 | ||
AK134028.1 | - | 3, 4 | 366 | |||
AK170351.1 | - | 3, 4 | 667 |
Gene Symbol: Tnfsfm13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159503.2 | NP_001152975.1 | 10, 11 | 1213 | ||
GenBank mRNA | BC096441.1 | - | 10, 11 | 925 | 3482756 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
