siRNA Details
Gene Symbol: Rpf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042556.1 | NP_001036021.1 | 10 | 1216 | ||
NM_023323.3 | NP_075812.3 | 10 | 866 | |||
GenBank mRNA | AB041810.1 | - | 10 | 1246 | ||
AK010776.1 | - | 10 | 860 | |||
AK011644.1 | - | 10 | 844 | |||
AK012188.1 | - | 10 | 866 | |||
AK012641.1 | - | 10 | 845 | |||
AK013395.1 | - | 10 | 841 | |||
AK017678.1 | - | 10 | 837 | |||
AK020030.1 | - | 10 | 827 | |||
AK051135.1 | - | 10 | 1217 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
