siRNA Details
Gene Symbol: Npc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023409.4 | NP_075898.1 | 4 | 1339 | ||
GenBank mRNA | AK008603.1 | - | 4 | 1126 | ||
AK149805.1 | - | 4 | 1130 | |||
AK151067.1 | - | 4 | 1126 | |||
AK153020.1 | - | 4 | 1126 | |||
AK154838.1 | - | 4 | 1126 | |||
AK158624.1 | - | 4 | 1307 | |||
AK167869.1 | - | 4 | 1127 | |||
AK170327.1 | - | 4 | 1127 | |||
BC003471.1 | - | 4 | 1141 | 2649564 |

More Information
Other Species Targeted

Gene Ontology Categories:
