siRNA Details
Gene Symbol: Sync | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023485.3 | NP_075974.3 | 4, 5 | 1528 | ||
GenBank mRNA | AJ251641.1 | - | 4, 5 | 1464 | ||
AK081983.1 | - | 4, 5 | 1533 | |||
AK166916.1 | - | 3, 4 | 1470 | |||
AK216975.1 | - | 1, 2 | 70 | |||
BC106855.2 | - | 4, 5 | 1445 | 40046886 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
