siRNA Details
Gene Symbol: Ddi2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017966.2 | NP_001017966.1 | 10 | 1814 | ||
GenBank mRNA | AK129015.1 | - | 1793 | |||
BC021415.1 | - | 10 | 1795 | |||
BC031491.1 | - | 3 | 568 |
Gene Symbol: Rsc1a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023544.5 | NP_076033.4 | 1 | 429 | ||
GenBank mRNA | AI854132.1 | - | 1 | 476 | ||
BC150752.1 | - | 1 | 429 | 9087663 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
