siRNA Details
Gene Symbol: Nfatc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168346.1 | NP_001161818.1 | 2 | 646 | ||
NM_023699.3 | NP_076188.3 | 2 | 646 | |||
GenBank mRNA | AK159078.1 | - | 2 | 492 | ||
EU887656.1 | - | 2 | 404 | |||
EU887657.1 | - | 2 | 404 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
