siRNA Details
Gene Symbol: Trp53rk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023815.4 | NP_076304.2 | 2 | 598 | ||
GenBank mRNA | AB028045.1 | - | 2 | 588 | ||
AK028411.1 | - | 2 | 599 | |||
AK046261.1 | - | 2 | 576 | |||
AK049291.1 | - | 2 | 583 | |||
AK160279.1 | - | 2 | 578 | |||
BC017155.1 | - | 2 | 616 | 4910610 |
Gene Symbol: 2810408M09Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007581.1 | NP_001007582.1 | 2 | 564 | ||
GenBank mRNA | AK013049.1 | - | 2 | 574 | ||
AK077144.1 | - | 2 | 572 | |||
AK167662.1 | - | 2 | 572 | |||
BC085263.1 | - | 2 | 564 | 6395509 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
