siRNA Details
Gene Symbol: Muc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023566.3 | NP_076055.3 | 4673 | |||
GenBank mRNA | BC034197.1 | - | 1, 2 | 526 | ||
BC036168.1 | - | 1, 2 | 124 | |||
BC036170.1 | - | 1, 2 | 494 | |||
BC039284.1 | - | 1, 2 | 455 | |||
BC039285.1 | - | 1, 2 | 340 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
