siRNA Details
Gene Symbol: Rab27a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB232622.1 | - | 2, 3 | 242 | ||
AF304376.1 | - | 3, 4 | 421 | |||
BB857060.1 | - | 3, 4 | 484 | |||
BC008173.1 | - | 4, 5 | 572 | 3710213 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
