siRNA Details
Gene Symbol: SEH1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031216.3 | NP_112493.2 | 9 | 1537 | ||
XM_011525744.1 | XP_011524046.1 | 10 | 1593 | |||
XM_017026025.1 | XP_016881514.1 | 8 | 1386 | |||
XM_017026027.1 | XP_016881516.1 | 9 | 1707 | |||
GenBank mRNA | AF136976.1 | - | 9 | 1506 | ||
AF255625.1 | - | 9 | 1441 | |||
AK226075.1 | - | 6 | 3927 | |||
BC012430.1 | - | 9 | 1491 | 3865994 | ||
BX538037.1 | - | 4 | 3246 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
