siRNA Details
Gene Symbol: Trp53rk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023815.4 | NP_076304.2 | 2 | 820 | ||
GenBank mRNA | AB028045.1 | - | 2 | 810 | ||
AK011574.2 | - | 2 | 781 | |||
AK028411.1 | - | 2 | 821 | |||
AK046261.1 | - | 2 | 798 | |||
AK049291.1 | - | 2 | 805 | |||
AK160279.1 | - | 2 | 800 | |||
BC017155.1 | - | 2 | 838 | 4910610 |
Gene Symbol: 2810408M09Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007581.1 | NP_001007582.1 | 2 | 786 | ||
GenBank mRNA | AK013049.1 | - | 2 | 796 | ||
AK167662.1 | - | 2 | 794 | |||
BC085263.1 | - | 2 | 786 | 6395509 |

More Information
Other Species Targeted

Gene Ontology Categories:
