siRNA Details
Gene Symbol: Ankrd36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023816.2 | NP_076305.2 | 1, 2 | 454 | ||
GenBank mRNA | AF294328.1 | - | 1, 2 | 456 | ||
AK005925.1 | - | 1, 2 | 428 | |||
AK132751.1 | - | 1, 2 | 455 | |||
BC120815.1 | - | 1, 2 | 232 | |||
BC120817.1 | - | 1, 2 | 229 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
