siRNA Details
Gene Symbol: Ush1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023649.2 | NP_076138.2 | 15, 16 | 1306 | ||
GenBank mRNA | AF228924.1 | - | 15, 16 | 1306 | ||
BC010819.1 | - | 15, 16 | 1308 | 4217086 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
