siRNA Details
Gene Symbol: 9530002B09Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023865.3 | NP_076354.1 | 5 | 797 | ||
GenBank mRNA | AF319955.1 | - | 5 | 782 | ||
AK020534.1 | - | 5 | 747 | |||
AY077734.1 | - | 5 | 776 |
