siRNA Details
Gene Symbol: Sapcd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023893.4 | NP_076382.3 | 2 | 254 | ||
GenBank mRNA | AK019275.1 | - | 2 | 155 | ||
BC147772.1 | - | 2 | 254 | 9087495 | ||
BC147780.1 | - | 2 | 254 | 9087500 | ||
BC172157.1 | - | 2 | 258 | |||
CF739520.1 | - | 2 | 152 |

More Information
Other Species Targeted

Gene Ontology Categories:
