siRNA Details
Gene Symbol: Chchd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024166.6 | NP_077128.2 | 4 | 827 | ||
GenBank mRNA | AK003399.1 | - | 4 | 662 | ||
AK012691.1 | - | 3 | 653 | |||
AK145492.1 | - | 4 | 645 | |||
AK160762.1 | - | 4 | 646 | |||
AK160779.1 | - | 4 | 647 | |||
AK162133.1 | - | 4 | 645 | |||
AK166796.1 | - | 4 | 647 | |||
AK167593.1 | - | 4 | 647 | |||
AK211922.1 | - | 1 | 176 | |||
AK216042.1 | - | 2 | 175 | |||
BC037682.1 | - | 3 | 2007 | |||
BC042717.1 | - | 4 | 634 | 3154544 | ||
BC069887.1 | - | 4 | 409 | |||
BC082287.1 | - | 4 | 637 | 6397957 | ||
BC083126.1 | - | 3 | 1970 | |||
BC084682.1 | - | 4 | 616 | 6530688 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
