siRNA Details
Gene Symbol: Cxx1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024170.2 | NP_077132.1 | 1 | 829 | ||
GenBank mRNA | AK002323.1 | - | 1 | 765 | ||
BC003436.1 | - | 1 | 748 | 2647372 |
Gene Symbol: Cxx1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001018063.1 | NP_001018073.1 | 1 | 797 | ||
GenBank mRNA | AK003646.1 | - | 1 | 776 | ||
AK206042.1 | - | 1 | 165 | |||
BC029838.1 | - | 1 | 797 | 3967024 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
