siRNA Details
Gene Symbol: Ndufa10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024197.1 | NP_077159.1 | 2 | 254 | ||
GenBank mRNA | AK005339.1 | - | 2 | 290 | ||
AK013675.1 | - | 2 | 298 | |||
AK046292.1 | - | 158 | ||||
AK145979.1 | - | 2 | 310 | |||
AK177419.1 | - | 1 | 108 | |||
AK184477.1 | - | 1 | 108 | |||
BC003439.1 | - | 2 | 254 | 2647815 |
