siRNA Details
Gene Symbol: Stambp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024239.2 | NP_077201.1 | 2 | 276 | ||
GenBank mRNA | AB010123.1 | - | 2 | 223 | ||
AK017600.1 | - | 2 | 231 | |||
AK019907.1 | - | 2 | 226 | |||
AK046995.1 | - | 2 | 228 | |||
AK136961.1 | - | 2 | 827 | |||
AK139391.1 | - | 2 | 251 | |||
AK168810.1 | - | 2 | 249 | |||
AK220541.1 | - | 2 | 199 | |||
BC003497.1 | - | 2 | 219 | 3153829 | ||
BC006939.1 | - | 2 | 274 | 2649718 | ||
BC025111.1 | - | 2 | 218 | 4194332 | ||
CJ074803.1 | - | 2 | 276 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_138531, XM_008763048, XM_008763045, XM_006236726, XM_008763046, and others.
Gene Ontology Categories:
