siRNA Details
Gene Symbol: ADGRF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153840.3 | NP_722582.2 | 11, 12 | 2714 | ||
XM_011514469.1 | XP_011512771.1 | 8, 9 | 2370 | |||
XM_017010735.1 | XP_016866224.1 | 8, 9 | 2472 | |||
XM_017010736.1 | XP_016866225.1 | 9, 10 | 2447 | |||
GenBank mRNA | AB045123.1 | - | 11, 12 | 2668 | ||
AY140952.1 | - | 5, 6 | 1907 | |||
CR627234.1 | - | 9, 10 | 8763 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
