siRNA Details
Gene Symbol: ACADVL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000018.3 | NP_000009.1 | 3, 4 | 363 | ||
NM_001270447.1 | NP_001257376.1 | 4, 5 | 299 | |||
NM_001270448.1 | NP_001257377.1 | 2, 3 | 438 | |||
XM_006721516.2 | XP_006721579.2 | 3, 4 | 316 | |||
XM_011523829.1 | XP_011522131.1 | 3, 4 | 316 | |||
XM_011523830.1 | XP_011522132.1 | 3, 4 | 316 | |||
GenBank mRNA | AK056083.1 | - | 2, 3 | 331 | ||
AK058109.1 | - | 3, 4 | 245 | |||
AK097243.1 | - | 3, 4 | 319 | |||
AK222518.1 | - | 3, 4 | 245 | |||
AK293549.1 | - | 4, 5 | 299 | |||
AK302653.1 | - | 3, 4 | 309 | |||
AL536186.3 | - | 3, 4 | 389 | |||
BC000399.2 | - | 3, 4 | 301 | 2822593 | ||
BC012912.1 | - | 3, 4 | 285 | 4471040 | ||
D43682.1 | - | 3, 4 | 242 | |||
DA945553.1 | - | 3, 4 | 332 | |||
X86556.1 | - | 3, 4 | 294 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
