siRNA Details
Gene Symbol: Ralgps2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159965.1 | NP_001153437.1 | 14, 15 | 1497 | ||
NM_001159966.1 | NP_001153438.1 | 14, 15 | 1395 | |||
NM_001159968.1 | NP_001153440.1 | 13, 14 | 1392 | |||
NM_023884.4 | NP_076373.3 | 14, 15 | 1437 | |||
GenBank mRNA | AF312924.1 | - | 13, 14 | 1345 | ||
AK018622.1 | - | 14, 15 | 1439 | |||
AK019543.1 | - | 13, 14 | 1395 | |||
AK029049.1 | - | 14, 15 | 1438 | |||
AK036803.1 | - | 13, 14 | 1369 | |||
AK165002.1 | - | 14, 15 | 1389 | |||
BC052663.1 | - | 14, 15 | 1414 | 30051209 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
