siRNA Details
Gene Symbol: Sapcd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023893.4 | NP_076382.3 | 3 | 429 | ||
GenBank mRNA | AK019275.1 | - | 3 | 330 | ||
BC147772.1 | - | 3 | 429 | 9087495 | ||
BC147780.1 | - | 3 | 429 | 9087500 | ||
BC172157.1 | - | 3 | 433 | |||
CF739520.1 | - | 3 | 327 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
