siRNA Details
Gene Symbol: Rps23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024175.3 | NP_077137.1 | 3, 4 | 367 | ||
GenBank mRNA | AK002573.1 | - | 3, 4 | 325 | ||
AK010594.1 | - | 3, 4 | 325 | |||
AK010608.1 | - | 3, 4 | 323 | |||
AK010667.1 | - | 3, 4 | 337 | |||
AK012287.1 | - | 3, 4 | 325 | |||
AK012438.1 | - | 3, 4 | 319 | |||
AK012574.1 | - | 3, 4 | 325 | |||
AK013720.1 | - | 3, 4 | 326 | |||
AK050580.1 | - | 3, 4 | 323 | |||
AK088085.1 | - | 3, 4 | 325 | |||
AK166994.1 | - | 3, 4 | 359 | |||
AK168587.1 | - | 3, 4 | 319 | |||
BC002145.1 | - | 3, 4 | 314 | 3484753 | ||
BC054435.1 | - | 2, 3 | 304 | 5373967 | ||
BC078418.1 | - | 2, 3 | 304 | 5329878 | ||
BC100603.1 | - | 3, 4 | 326 | 6760902 | ||
BQ746768.1 | - | 3, 4 | 305 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
