siRNA Details
Gene Symbol: Ndufa10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024197.1 | NP_077159.1 | 10 | 1184 | ||
GenBank mRNA | AK013675.1 | - | 10 | 1227 | ||
AK046292.1 | - | 1088 | ||||
AK145979.1 | - | 10 | 1240 | |||
AK203226.1 | - | 2 | 188 | |||
BC003439.1 | - | 10 | 1184 | 2647815 |
