siRNA Details
Gene Symbol: Stt3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK145674.1 | - | 16 | 3953 | ||
AK148065.1 | - | 16 | 4010 | |||
BC013054.1 | - | 6 | 2428 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
