siRNA Details
Gene Symbol: Fam13c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143776.1 | NP_001137248.1 | 14 | 2896 | ||
NM_001143777.1 | NP_001137249.1 | 15 | 3003 | |||
NM_024244.4 | NP_077206.3 | 14 | 2899 | |||
GenBank mRNA | AK004797.1 | - | 14 | 2883 | ||
AK034001.1 | - | 1 | 686 | |||
AK041147.1 | - | 14 | 2897 | |||
AK220421.1 | - | 14 | 4441 | |||
BC003231.1 | - | 3 | 1398 | |||
BC066835.1 | - | 3 | 1351 | |||
BC129839.1 | - | 14 | 2852 | 40060810 | ||
BC129840.1 | - | 14 | 2855 | 40060811 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
