siRNA Details
Gene Symbol: Desi2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024282.3 | NP_077244.1 | 3 | 679 | ||
GenBank mRNA | AK020014.1 | - | 3 | 564 | ||
AK029574.1 | - | 4 | 753 | |||
AK049579.1 | - | 3 | 448 | |||
AK146492.1 | - | 3 | 412 | |||
AK160733.1 | - | 3 | 268 | |||
AK216786.1 | - | 2 | 151 | |||
BC002200.1 | - | 3 | 348 | 3489351 | ||
BC046816.1 | - | 3 | 386 | 5702476 |
