siRNA Details
Gene Symbol: Ky | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024291.3 | NP_077253.3 | 1, 2 | 146 | ||
GenBank mRNA | AJ293727.2 | - | 1, 2 | 168 | ||
AK047076.1 | - | 1, 2 | 537 |

More Information
Other Species Targeted

Gene Ontology Categories:
