siRNA Details
Gene Symbol: Tnfrsf22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023680.3 | NP_076169.2 | 7 | 1051 | ||
GenBank mRNA | AK012838.1 | - | 7 | 1052 | ||
AY046551.1 | - | 6 | 1005 | |||
BC076592.1 | - | 6 | 1020 |
Gene Symbol: Tnfrsf23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK154786.1 | - | 6 | 951 | ||
AK170037.1 | - | 6 | 1026 | |||
AY165625.1 | - | 6 | 940 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
