siRNA Details
Gene Symbol: BC005537 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK050530.1 | - | 1992 | |||
AK088171.1 | - | 474 | ||||
AK160094.1 | - | 2154 | ||||
BC002217.1 | - | 1571 | 3490515 | |||
BC005537.1 | - | 2834 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
