siRNA Details
Gene Symbol: Nrbf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001036293.2 | NP_001031370.1 | 2 | 250 | ||
GenBank mRNA | AK002810.1 | - | 2 | 252 | ||
AK004200.1 | - | 2 | 172 | |||
AK181810.1 | - | 2 | 103 | |||
AK189272.1 | - | 2 | 103 | |||
AK205591.1 | - | 2 | 130 | |||
BC019448.1 | - | 2 | 250 | 4212132 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
