siRNA Details
Gene Symbol: Tmem254c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270498.1 | NP_001257427.1 | 481 | |||
NM_001270499.1 | NP_001257428.1 | 301 | ||||
GenBank mRNA | BY703810.1 | - | 221 | |||
CK128852.1 | - | 396 | ||||
CX201482.1 | - | 301 |
Gene Symbol: Tmem254a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025311.3 | NP_079587.1 | 312 | |||
NM_026679.2 | NP_080955.1 | 481 | ||||
GenBank mRNA | AK002342.1 | - | 2, 3 | 300 | ||
AK002441.1 | - | 2, 3 | 412 | |||
AK003554.1 | - | 2, 3 | 221 | |||
AK008299.1 | - | 2, 3 | 272 | |||
BC031854.1 | - | 2, 3 | 365 | 4194400 | ||
BY270140.1 | - | 313 |
Gene Symbol: Tmem254b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270495.1 | NP_001257424.1 | 2, 3 | 465 | ||
NM_001270496.1 | NP_001257425.1 | 2, 3 | 223 | |||
GenBank mRNA | BY713360.1 | - | 2, 3 | 465 | ||
CK793021.1 | - | 2, 3 | 221 | |||
DV046977.1 | - | 1, 2 | 83 |
