siRNA Details
Gene Symbol: Tceal3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029978.2 | NP_001025149.2 | 3 | 438 | ||
GenBank mRNA | AK003156.1 | - | 3 | 354 | ||
AK017585.1 | - | 3 | 354 | |||
AK158996.1 | - | 3 | 474 | |||
BC027131.1 | - | 3 | 361 | 5358255 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
