siRNA Details
Gene Symbol: Tmem254a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025311.3 | NP_079587.1 | 752 | |||
NM_026679.2 | NP_080955.1 | 921 | ||||
GenBank mRNA | AK002342.1 | - | 4 | 740 | ||
AK002441.1 | - | 4 | 852 | |||
AK003554.1 | - | 4 | 661 | |||
AK008299.1 | - | 4 | 712 | |||
BC031854.1 | - | 4 | 805 | 4194400 |
Gene Symbol: Tmem254b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270495.1 | NP_001257424.1 | 4 | 905 | ||
NM_001270496.1 | NP_001257425.1 | 4 | 663 | |||
GenBank mRNA | CK793021.1 | - | 4 | 661 | ||
DV046977.1 | - | 3 | 523 |
Gene Symbol: Tmem254c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270498.1 | NP_001257427.1 | 921 | |||
NM_001270499.1 | NP_001257428.1 | 741 | ||||
GenBank mRNA | BY703810.1 | - | 661 |
