siRNA Details
Gene Symbol: Tmem176a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098271.1 | NP_001091741.1 | 6, 7 | 756 | ||
NM_025326.4 | NP_079602.4 | 6, 7 | 808 | |||
GenBank mRNA | AB063313.1 | - | 5, 6 | 698 | ||
AK002546.1 | - | 6, 7 | 784 | |||
AK004315.1 | - | 6, 7 | 782 | |||
AK079378.1 | - | 770 | ||||
AK150576.1 | - | 6, 7 | 772 | |||
AK162991.1 | - | 4, 5 | 2016 | |||
AK165660.1 | - | 6, 7 | 782 | |||
BC010831.1 | - | 6, 7 | 752 | 4221626 | ||
CT010190.1 | - | 5, 6 | 698 |
