siRNA Details
Gene Symbol: Tceal5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177919.2 | NP_808587.1 | 3 | 565 | ||
GenBank mRNA | AK032140.1 | - | 3 | 567 | ||
BC100415.1 | - | 3 | 590 | 6535676 | ||
BC115460.1 | - | 2 | 476 | 40100235 | ||
BC115461.1 | - | 2 | 476 | 40100237 |
Gene Symbol: Tceal3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029978.2 | NP_001025149.2 | 3 | 660 | ||
GenBank mRNA | AK003156.1 | - | 3 | 576 | ||
AK017585.1 | - | 3 | 576 | |||
AK158996.1 | - | 3 | 696 | |||
BC027131.1 | - | 3 | 583 | 5358255 |
Gene Symbol: Tceal6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025355.4 | NP_079631.2 | 3 | 642 | ||
GenBank mRNA | AK005297.1 | - | 3 | 644 | ||
BC087890.1 | - | 3 | 642 | 6533845 |

More Information
Other Species Targeted

Gene Ontology Categories:
