siRNA Details
Gene Symbol: Tmem57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025382.6 | NP_079658.2 | 10 | 1966 | ||
GenBank mRNA | AK003528.1 | - | 4 | 482 | ||
AK014449.1 | - | 2 | 220 | |||
AY364165.1 | - | 10 | 1751 | |||
AY846873.1 | - | 10 | 1967 | |||
AY846874.1 | - | 10 | 1967 | |||
BC043306.1 | - | 7 | 1398 | |||
BC054769.1 | - | 10 | 1859 | 6415086 | ||
BC056944.1 | - | 3 | 320 |
