siRNA Details
Gene Symbol: Nrbf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001036293.2 | NP_001031370.1 | 4 | 1010 | ||
GenBank mRNA | AK002810.1 | - | 4 | 1009 | ||
BC019448.1 | - | 4 | 1010 | 4212132 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
