siRNA Details
Gene Symbol: Tomm6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164729.1 | NP_001158201.1 | 3 | 966 | ||
NM_025365.3 | NP_079641.1 | 3 | 912 | |||
GenBank mRNA | AK010470.1 | - | 3 | 624 | ||
CA321392.1 | - | 3 | 496 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
