siRNA Details
Gene Symbol: Mrps36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190264.1 | NP_001177193.1 | 4 | 454 | ||
NM_025369.3 | NP_079645.1 | 4 | 460 | |||
GenBank mRNA | AK003111.1 | - | 4 | 390 | ||
AK003765.1 | - | 4 | 437 | |||
AK007512.1 | - | 4 | 431 | |||
AK009050.1 | - | 4 | 439 | |||
AK009966.1 | - | 4 | 385 | |||
BC028499.1 | - | 4 | 416 | 1382215 | ||
BC096576.1 | - | 4 | 425 | 1401014 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
