siRNA Details
Gene Symbol: Tmed6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025458.2 | NP_079734.1 | 1 | 192 | ||
GenBank mRNA | AK007320.1 | - | 1 | 188 | ||
AK007518.1 | - | 1 | 193 | |||
AK007830.1 | - | 1 | 186 | |||
AK007885.1 | - | 1 | 193 | |||
BC086923.1 | - | 1 | 192 | 6760985 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
