siRNA Details
Gene Symbol: Fam134b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034851.2 | NP_001030023.1 | 7 | 837 | ||
NM_001277315.1 | NP_001264244.1 | 5 | 926 | |||
NM_001277316.1 | NP_001264245.1 | 5 | 509 | |||
NM_001277317.1 | NP_001264246.1 | 5 | 487 | |||
NM_001277318.1 | NP_001264247.1 | 5 | 459 | |||
NM_025459.3 | NP_079735.2 | 6 | 627 | |||
GenBank mRNA | AK007506.1 | - | 5 | 488 | ||
AK015182.1 | - | 6 | 449 | |||
AK146972.1 | - | 5 | 510 | |||
AK167327.1 | - | 5 | 927 | |||
BC019494.1 | - | 6 | 566 | 4460992 | ||
BC054415.1 | - | 5 | 446 | 3487542 |
