siRNA Details
Gene Symbol: 1700011A15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025487.3 | NP_079763.1 | 5 | 874 | ||
GenBank mRNA | AK007046.1 | - | 5 | 844 | ||
BC048463.1 | - | 5 | 851 | 6742420 |

More Information
Other Species Targeted

Gene Ontology Categories:
