siRNA Details
Gene Symbol: 1700018B24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK006080.1 | - | 1 | 319 | ||
AK006613.1 | - | 1 | 319 | |||
AK018885.1 | - | 1 | 288 | |||
BC049578.1 | - | 1 | 302 | |||
BC061086.1 | - | 1 | 318 |

More Information
Other Species Targeted

