siRNA Details
Gene Symbol: Thoc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285780.1 | NP_001272709.1 | 7 | 616 | ||
NM_025435.3 | NP_079711.1 | 6 | 538 | |||
GenBank mRNA | AK005176.1 | - | 6 | 465 | ||
AK007440.1 | - | 6 | 444 | |||
AK010958.1 | - | 6 | 479 | |||
AK147003.1 | - | 7 | 519 | |||
BC027113.1 | - | 5 | 1486 | |||
BC042209.1 | - | 6 | 465 | |||
BC054419.1 | - | 6 | 498 | 4010343 | ||
BC116919.1 | - | 6 | 503 | 40126238 | ||
BC116945.1 | - | 6 | 503 | 40126264 | ||
CK329337.1 | - | 6 | 539 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
